Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucin, MP Biomedicals™

Catalog No. ICN15574291 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15574291 1 g
ICN15574280 100 mg
ICN15574290 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15574291 Supplier MP Biomedicals, Inc Supplier No. 0215574291
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84195-52-8

Specifications

Color Tan
Quantity 1 g
Formula Weight ≈400,0002
Physical Form Flaky Powder
Chemical Name or Material Mucin
EINECSNumber 282-357-4
Recommended Storage Store at 4°C, dessicate.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.