Learn More
Newcomer Supply Mucin Mucicarmine Control Slides, 98/set
SDP
Catalog No. NC1954634
Encompass
Newcomer Supply Mucin Mucicarmine Control Slides are for the positive histochemical staining of acid epithelial mucins (sialomucin, sulfomucin). Mucin Mucicarmine Control Slides contain sections of positive staining colon.
Catalog No. NC1954634 Supplier Newcomer Supply Supplier No. 4455B
Newcomer Supply Mucin Mucicarmine Control Slides are for the positive histochemical staining of acid epithelial mucins (sialomucin, sulfomucin). Mucin Mucicarmine Control Slides contain sections of positive staining colon.