Learn More
Newcomer Supply Mucin, Mayer Mucicarmine Stain Kit, 500 ml kit
SDP
Supplier:  Newcomer Supply 9151B
Encompass
Newcomer Supply Mucin, Mayer Mucicarmine Stain Kit procedure, with included microwave modification, is used to stain acid epithelial mucin (sialomucin, sulfomucin) and is also useful for the demonstration of the encapsulated yeast Cryptococcus neoformans.
Catalog No. NC2102280