Learn More
ACROBiosystems Recombinant Protein;Human Mucin-1 / MUC-1 Protein, Fc Tag;HEK293;1MG;MU1,Mucin 1,MUC1,CD227,EMA,H23AG,KL-6,MAM6,MUC-1,MUC-1/SEC,MUC-1/X,MUC1/ZD,PEM,PEMT,PUM,CA 15-3,Episialin
Supplier:  ACROBiosystems MU1H52521MG
Encompass_Preferred
Recombinant Protein;Human Mucin-1 / MUC-1 Protein, Fc Tag;1MG;Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession # AAI20976).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.8 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8021