Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

muco-Inositol 98.0+%, TCI America™
SDP

Catalog No. I0630100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0630100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0630100MG Supplier TCI America Supplier No. I0630100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41546-34-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD01321249
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 286°C
Color White
Quantity 100 mg
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material muco-Inositol
EINECSNumber (3)-3265
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.