Learn More
MOLTOX Molecular Toxicology Inc Mueller Hinton Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948832
Encompass_Preferred
Mueller Hinton Agar is used in antimicrobial disc susceptibility testing as well as for cultivating a wide variety of microorganisms. This media is designed to contain low levels of thymine and thymidine as well as controlled levels of calcium and magnesium. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-832 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S055
Mueller Hinton Agar is used in antimicrobial disc susceptibility testing as well as for cultivating a wide variety of microorganisms. This media is designed to contain low levels of thymine and thymidine as well as controlled levels of calcium and magnesium. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.