Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Mueller Hinton Agar
SDP

Catalog No. 23001799 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-799 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-799 Supplier Hardy Diagnostics™ Supplier No. G45
Add to Cart
Add to Cart
This item is not returnable. View return policy

For antimicrobial susceptibility testing of non-fastidious bacteria by utilizing disk diffusion sensitivity method

  • Polystyrene Petri plates feature raised rings for stacking ability and prevent sliding
  • Packaged in sleeves of 10 plates in a breathable celluose bag that prevents build-up of condensation and excess moisture

Specifications

Appearance Translucent
Certifications/Compliance CLSI M100, ISO 16782
Color Amber
Content And Storage 2°C to 8°C
Form Prepared Medium
Format Plate
Media Type Mueller Hinton Agar
Product Type Agar Plate
Target Organism Class Non-fastidious, Gram-positive Organisms, Non-fastidious, Gram-negative Organisms
Test Type Antimicrobial Susceptibility
Fill Volume 18 mL
Formulation Acid Hydrolysate of Casein - 17.5gm, Beef Extract - 2.0gm, Starch - 1.5gm, Agar - 17.0gm
For Use With (Application) Antimicrobial Susceptibility Testing, Disk Diffusion Sensitivity Method (Kirby-Bauer)
Incubation Conditions 35°C
Incubation Time 24 hr.
Material Polystyrene
Packaging Type Bag
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 100 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Escherichia coli, Staphylococcus species, Enterococcus faecalis, Pseudomonas protegens
Protein Source Beef, Casein
Source Beef, Casein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.