Learn More
Bioworld Mueller Hinton Agar, 500 g
Catalog No. 50254336
Encompass_Preferred
Mueller Hinton AgarFor antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method, as standardized by the National Committee for Clinical Laboratory Standards (NCCLS)pH: 7.3Working Concentration: 38 g/LDehydrated Appearance: beige, homogeneous powderPrepared Appearance: light to medium amber agarSoluble in boiling waterContains (g/L):Beef Extrac: 2g Acid Hydrolysate of Casein: 17.5g Starch: 1.5g Agar: 17g
Catalog No. 50-254-336 Supplier Bioworld Supplier No. 306200541
Mueller Hinton AgarFor antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method, as standardized by the National Committee for Clinical Laboratory Standards (NCCLS)pH: 7.3Working Concentration: 38 g/LDehydrated Appearance: beige, homogeneous powderPrepared Appearance: light to medium amber agarSoluble in boiling waterContains (g/L):Beef Extrac: 2g Acid Hydrolysate of Casein: 17.5g Starch: 1.5g Agar: 17g