Learn More
Alkali Scientific Mueller Hinton Agar 75mL Monoplate, 10/Pk
SDP
Catalog No. 5000662164
Encompass_Preferred
Mueller Hinton Agar, 75mL Monoplate, 10/Pk is a non selective culture medium widely used for antimicrobial susceptibility testing using standardized disk diffusion methods such as the Kirby Bauer technique. The deep 75mL fill volume ensures consistent antibiotic diffusion across the agar surface, allowing accurate and reproducible zone of inhibition measurements. This formulation supports the growth of a wide range of non fastidious bacterial species and meets the performance requirements recommended by CLSI guidelines. The plates are supplied in mono wrapped packaging to maintain sterility and reduce contamination risk during laboratory workflows. Mueller Hinton Agar is the industry standard medium used in clinical microbiology, pharmaceutical research, and antimicrobial resistance testing programs.
Catalog No. 50-006-62164 Supplier Alkali Scientific Supplier No. MHAP01
Mueller Hinton Agar, 75mL Monoplate, 10/Pk is a non selective culture medium widely used for antimicrobial susceptibility testing using standardized disk diffusion methods such as the Kirby Bauer technique. The deep 75mL fill volume ensures consistent antibiotic diffusion across the agar surface, allowing accurate and reproducible zone of inhibition measurements. This formulation supports the growth of a wide range of non fastidious bacterial species and meets the performance requirements recommended by CLSI guidelines. The plates are supplied in mono wrapped packaging to maintain sterility and reduce contamination risk during laboratory workflows. Mueller Hinton Agar is the industry standard medium used in clinical microbiology, pharmaceutical research, and antimicrobial resistance testing programs.