Learn More
HIMEDIA Mueller Hinton Agar, M173, Microbiology Culture Media, Dehydrated Culture Media, Agar, Antimicrobial Susceptibility Testing, CLSI Standardized, Powder, 500 g
Catalog No. NC4013700
Encompass_Preferred
Mueller Hinton Agar is the internationally referenced medium for antimicrobial susceptibility testing of rapidly growing aerobic bacteria from clinical isolates. Its transparent beef heart infusion and casein hydrolysate base produces sharp, easy-to-read zones of inhibition, and every lot is performance tested and standardized to current CLSI (formerly NCCLS) protocols for reproducible disc diffusion (Kirby-Bauer) results. The controlled formulation supports dependable zone and MIC measurement across a broad organism range, final pH 7.3. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013700 Supplier HIMEDIA Supplier No. M173500G
Mueller Hinton Agar is the internationally referenced medium for antimicrobial susceptibility testing of rapidly growing aerobic bacteria from clinical isolates. Its transparent beef heart infusion and casein hydrolysate base produces sharp, easy-to-read zones of inhibition, and every lot is performance tested and standardized to current CLSI (formerly NCCLS) protocols for reproducible disc diffusion (Kirby-Bauer) results. The controlled formulation supports dependable zone and MIC measurement across a broad organism range, final pH 7.3. Dehydrated powder, store in a cool dry place. Packaged as 500 g.