Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Mueller Hinton Agar (MHA)
SDP

Catalog No. 23001812 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-812 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-812 Supplier Hardy Diagnostics™ Supplier No. H11
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mueller Hinton Media is recommended for use in the cultivation of a wide variety of microorganisms. Mueller Hinton Agar can be used for Kirby Bauer disk diffusion sensitivity testing of non-fastidious organisms.

  • Mueller Hinton Broth is recommended for preparing suspensions of microorganisms for disk diffusion sensitivity testing.

  • Mueller Hinton Media contains beef infusion, casamino acids, and starch.
    • Starch acts as a colloid that protects against toxic material in the medium
    • Beef infusion and casamino acids provide energy and nutrients

Specifications

Appearance Translucent
Certifications/Compliance CLSI M100, ISO 16782
Color Light Amber
Content And Storage 2°C to 8°C
Form Agar
Format Plate
Media Type Mueller Hinton Agar
Product Type Agar Plate
Target Organism Class Non-fastidious, Gram-negative Organisms, Non-fastidious, Gram-positive Organisms
Test Type Antibiotic Susceptibility Sensitivity
Fill Volume 69 mL
Formulation Acid Hydrolysate of Casein - 17.5gm, Beef Extract - 2.0gm, Starch - 1.5gm, Agar - 17.0gm
For Use With (Application) Antimicrobial Susceptibility Testing, Cultivation of Microorganisms
Incubation Conditions 35°C
Incubation Time 24 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Nylon Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 150 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 55 mm
Species Escherichia coli, Enterococcus faecalis, Pseudomonas species
Protein Source Casein, Beef
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.