Learn More
HIMEDIA Mueller Hinton Agar No. 2, M1084, Microbiology Culture Media, Dehydrated Culture Media, Agar, Antimicrobial Susceptibility Testing, Bauer-Kirby Method, Powder, 500 g
Catalog No. NC4013672
Encompass_Preferred
Mueller Hinton Agar No. 2 is formulated for testing the susceptibility of common, rapidly growing bacteria with antimicrobial discs by the Bauer-Kirby method. It is manufactured to contain low levels of thymine, thymidine, calcium and magnesium, the variables most likely to distort zone sizes, ensuring accurate and reproducible results. Beef heart infusion, casein hydrolysate, starch and agar provide a transparent base at final pH 7.3. A dependable choice for routine and reference susceptibility testing. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013672 Supplier HIMEDIA Supplier No. M1084500G
Mueller Hinton Agar No. 2 is formulated for testing the susceptibility of common, rapidly growing bacteria with antimicrobial discs by the Bauer-Kirby method. It is manufactured to contain low levels of thymine, thymidine, calcium and magnesium, the variables most likely to distort zone sizes, ensuring accurate and reproducible results. Beef heart infusion, casein hydrolysate, starch and agar provide a transparent base at final pH 7.3. A dependable choice for routine and reference susceptibility testing. Dehydrated powder, store in a cool dry place. Packaged as 500 g.