Learn More
Teknova Mueller Hinton Agar Plates. 100X100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387798
Encompass
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.
Catalog No. 50-238-7798 Supplier Teknova Supplier No. M00294PK
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.