Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Mueller Hinton with Blood Agar
SDP

Catalog No. 23001768 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-768 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-768 Supplier Hardy Diagnostics™ Supplier No. H21
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mueller Hinton Agar with 5% Sheep Blood. For disk diffusion sensitivity testing of Streptococcus spp.

  • The basal medium for Mueller Hinton with Blood is Mueller Hinton Agar supplemented with 5% defibrinated sheep blood

  • Mueller Hinton Media contains beef infusion, casamino acids, and starch.
    • Starch acts as a colloid that protects against toxic material in the medium
    • Beef infusion and casamino acids provide energy and nutrients
  • Optically clear polystrene for distortion free viewing

Specifications

Appearance Opaque
Certifications/Compliance CLSI, ISO 13485
Color Cherry Red
Content And Storage 2°C to 8°C
Additive 5% Defibrinated sheep blood, Starch, Agar
Detects Streptococcus spp., Streptococcus pneumoniae
Format Plate
Media Type Agar
Product Type Agar Plate
Target Organism Class Fastidious Organisms
Test Type Antimicrobial Susceptibility
Fill Volume 72 mL
Formulation Acid Hydrolysate of Casein - 17.5gm, Beef Extract - 2.0gm, Starch - 1.5gm, Sheep Blood - 50.0ml, Agar - 17.0gm (per 950ml of deionized water)
For Use With (Application) Disk diffusion sensitivity testing
Incubation Conditions 35°C
Incubation Time 24 hr.
No. per Pack 10
Packaging Type Bag
Quantity 10 Plates
Dimensions (Dia. x H) 150 x 15 mm
Width (Metric) 150 mm
Height (Metric) 15 mm
Diameter (Metric) 150 mm
Species Streptococcus pneumoniae
Protein Source Beef infusion, Casamino acids, Acid Hydrolysate of Casein, Beef Extract
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.