Learn More
Teknova Mueller Hinton Broth, 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387282
Encompass
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.
Catalog No. 50-238-7282 Supplier Teknova Supplier No. M58992PK
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.