Learn More
MOLTOX Molecular Toxicology Inc Mueller Hinton II Broth (Cation Adjusted), 100 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948789
Encompass_Preferred
Mueller Hinton II Broth is used in susceptability testing of aerobic and facultatively anaerobic bacteria.This medium is cation-adjusted to calcium and magnesium ion concentrations. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-789 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26638
Mueller Hinton II Broth is used in susceptability testing of aerobic and facultatively anaerobic bacteria.This medium is cation-adjusted to calcium and magnesium ion concentrations. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.