Learn More
Teknova Mueller Hinton II Agar Plates. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388037
Encompass
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.
Catalog No. 50-238-8037 Supplier Teknova Supplier No. M0025
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.