Learn More
MOLTOX Molecular Toxicology Inc Mueller Hinton Broth w/ Cations, 500 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948781
Encompass_Preferred
Mueller Hinton Broth is used in antimicrobial susceptibility testing as well as for cultivating a wide variety of fastidious and nonfastidious microorganisms. This medium is not cation-adjusted. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-781 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26617.047
Mueller Hinton Broth is used in antimicrobial susceptibility testing as well as for cultivating a wide variety of fastidious and nonfastidious microorganisms. This medium is not cation-adjusted. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.