Learn More
Research Products International Corp MUG [4-Methylumbelliferyl-B-D-glucuronide Trihydrate], 1 Gram
SDP
Catalog No. 50213482
Encompass_Preferred
Fluorescent substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene isolated from E. coli. Fluorescence assay allows quantitation of GUS activity in protein extracts at peak excitation of 365 nm (UV) and a peak emission of 455 nm (blue).
Catalog No. 50-213-482 Supplier Research Products International Corp Supplier No. M659001.0
Fluorescent substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene isolated from E. coli. Fluorescence assay allows quantitation of GUS activity in protein extracts at peak excitation of 365 nm (UV) and a peak emission of 455 nm (blue).