Learn More
ATCC MUG-Chor1; Chordoma; Human (Homo sapiens)
Catalog No. 502383586
Encompass
MUG-Chor1 is a mesenchymal like cell line that was isolated in sacrum from the 2009 of a 57-year-old, White female patient with chordoma. Chordoma is a rare slow-growing tumor type and MUG-Chor1 is a relatively slow-growing cell line. MUG-Chor1 has a heterogeneous morphology consisting of physaliferous cells with mucinous intercellular substance, which represent typical chordoma features. The cells contain amplification of transcription factor T (Brachyury) that is most specific marker for chordoma, as well as loss of PTEN. This cell line was accessioned with the support of the Chordoma Foundation, a nonprofit organization working to improve the lives of chordoma patients by accelerating research to develop effective treatments for chordoma. This cell line was deposited by Beate Rinner and Bernadette Liegl.
Catalog No. 50-238-3586 Supplier ATCC Supplier No. CRL3219
MUG-Chor1 is a mesenchymal like cell line that was isolated in sacrum from the 2009 of a 57-year-old, White female patient with chordoma. Chordoma is a rare slow-growing tumor type and MUG-Chor1 is a relatively slow-growing cell line. MUG-Chor1 has a heterogeneous morphology consisting of physaliferous cells with mucinous intercellular substance, which represent typical chordoma features. The cells contain amplification of transcription factor T (Brachyury) that is most specific marker for chordoma, as well as loss of PTEN. This cell line was accessioned with the support of the Chordoma Foundation, a nonprofit organization working to improve the lives of chordoma patients by accelerating research to develop effective treatments for chordoma. This cell line was deposited by Beate Rinner and Bernadette Liegl.