Learn More
TARGETMOL CHEMICALS INC MULBERRIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ01615MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mulberrin (Kuwanon C) is an inhibitor of OATP2B1-mediated estrone-3-sulfate (E3S) uptake (IC50 1.8 ±1.5 uM). purity: 99%
Catalog No. 50-257-8821