Learn More
Medchemexpress LLC HY-N3513 5mg Medchemexpress, Mulberrin CAS:62949-79-5 Purity:>98%
SDP
Catalog No. 501970414
Encompass_Preferred
Medchemexpress, HY-N3513 5mg Mulberrin CAS:62949-79-5 Mulberrin is a strong inhibitor of organic anion-transporting polypeptide 2B1 (OATP2B1)-mediated estrone-3-sulfate (E3S) uptake with an IC50 value being 1.8 ±1.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0414 Supplier Medchemexpress LLC Supplier No. HYN35135MG
Medchemexpress, HY-N3513 5mg Mulberrin CAS:62949-79-5 Mulberrin is a strong inhibitor of organic anion-transporting polypeptide 2B1 (OATP2B1)-mediated estrone-3-sulfate (E3S) uptake with an IC50 value being 1.8 ±1.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.