Learn More
TARGETMOL CHEMICALS INC Mulberrofuran Q 1MG
SDP
Catalog No. 502880278
Encompass_Preferred
Also available in 1 mL, 10 mg and bulk. Please contact Fisher for quotes. Mulberrofuran Q is an active compound that inhibits the formation of 12-hydroxy-5,8,10-heptadecatrienoic acid (HHT) and thromboxane B2 (cyclooxygenase product) , which protects neuronal cells from hypoxia-glycorrhoea-deficiency (OGD)-induced oxidative stress. Purity 98%
Catalog No. 50-288-0278 Supplier TARGETMOL CHEMICALS INC Supplier No. TN19571MG
Also available in 1 mL, 10 mg and bulk. Please contact Fisher for quotes. Mulberrofuran Q is an active compound that inhibits the formation of 12-hydroxy-5,8,10-heptadecatrienoic acid (HHT) and thromboxane B2 (cyclooxygenase product) , which protects neuronal cells from hypoxia-glycorrhoea-deficiency (OGD)-induced oxidative stress. Purity 98%