Learn More
Apexbio Technology LLC Mulberroside A 102841-42-9 20mg
SDP
Supplier:  Apexbio Technology LLC N152020
Encompass_Preferred
Mulberroside A (CAS 102841-42-9) is a natural phenolic compound isolated from species of the Moraceae family particularly Morus genus It is known to exert biological effects by modulating cellular oxidative stress and inhibiting melanin biosynthesis with studies indicating its regulatory role in tyrosinase activity Mulberroside A has been investigated for its antioxidant and anti-inflammatory properties as well as its potential to suppress melanogenesis in cellular models These activities make it a suitable molecule for research into dermatological disorders oxidative stress-related conditions and the mechanistic study of natural product-based therapeutics
Catalog No. 50-000-89628