Learn More
Medchemexpress LLC HY-N0619 5mg Medchemexpress, Mulberroside A CAS:102841-42-9 Purity:>98%
SDP
Catalog No. 501933644
Encompass_Preferred
Medchemexpress, HY-N0619 5mg Mulberroside A CAS:102841-42-9 Mulberroside A is one of the main bioactive constituent in mulberry (Morus alba L.) [1] . Mulberroside A decreases the expressions of TNF-α , IL-1β , and IL-6 and inhibits the activation of NALP3, caspase-1, and NF-κB and the phosphorylation of ERK, JNK, and p38, exhibiting anti-inflammatory antiapoptotic effects [2] . Mulberroside A shows inhibitory activity against mushroom tyrosinase with an IC 50 of 53.6 μM [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3644 Supplier Medchemexpress LLC Supplier No. HYN06195MG
Medchemexpress, HY-N0619 5mg Mulberroside A CAS:102841-42-9 Mulberroside A is one of the main bioactive constituent in mulberry (Morus alba L.) [1] . Mulberroside A decreases the expressions of TNF-α , IL-1β , and IL-6 and inhibits the activation of NALP3, caspase-1, and NF-κB and the phosphorylation of ERK, JNK, and p38, exhibiting anti-inflammatory antiapoptotic effects [2] . Mulberroside A shows inhibitory activity against mushroom tyrosinase with an IC 50 of 53.6 μM [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.