Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mulberroside C | 102841-43-0 | MFCD20527296 | 99.8% | 458.46 | C24H26O9 | 10 MG
SDP

Catalog No. 5000296292
Encompass_Preferred

Mulberroside C is a natural stilbenoid glycoside isolated from mulberry (Morus alba L.) that has reported HCV replicon inhibitory and antiviral activity. It is provided as a high-purity research chemical suitable for biochemical and pharmacological studies; handle using appropriate laboratory safety procedures.

  • Reported HCV replicon inhibitor with antiviral activity.
  • High purity: 99.77% (as listed by supplier).
  • Chemical formula: C24H26O9; molecular weight: 458.46 g/mol.
  • CAS number: 102841-43-0 for compound identification.
  • Available in small research sizes such as 10 MG for assay development.
  • Storage: store at 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-96292 Supplier Medchemexpress LLC Supplier No. HYN062010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mulberroside C is a natural stilbenoid glycoside isolated from mulberry (Morus alba L.) that has reported HCV replicon inhibitory and antiviral activity. It is provided as a high-purity research chemical suitable for biochemical and pharmacological studies; handle using appropriate laboratory safety procedures.

  • Reported HCV replicon inhibitor with antiviral activity.
  • High purity: 99.77% (as listed by supplier).
  • Chemical formula: C24H26O9; molecular weight: 458.46 g/mol.
  • CAS number: 102841-43-0 for compound identification.
  • Available in small research sizes such as 10 MG for assay development.
  • Storage: store at 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.