Learn More
Cayman Chemical Phytol, Purity: ≥85% (mixture of isomers), CAS: 7541-49-3, 250 g
SDP
Catalog No. 502967862
Encompass_Preferred
A degradation product of chlorophyll that is converted to phytanic acid, which can accumulate to toxic levels in a number of metabolic disorders; activates RXRs (Kis range from 2.3-67.2 µM) and promotes the activity of PPAR/RXR heterodimers
Catalog No. 50-296-7862 Supplier Cayman Chemical Supplier No. 17401250 G
A degradation product of chlorophyll that is converted to phytanic acid, which can accumulate to toxic levels in a number of metabolic disorders; activates RXRs (Kis range from 2.3-67.2 µM) and promotes the activity of PPAR/RXR heterodimers