Learn More
Alkali Scientific ASI™ 96 Well Cell Culture TC Treated Plate, White, Sterile, 50/cs
SDP
Catalog No. 502335969
Encompass_Preferred
White plates for luminescence assays provide maximum reflection and help minimize cross-talk. The entire body and flat surface of this plate is homogenious white color. Provided with a compatible, low evaporation lid. ▪ HTS (High-Throughput Screening) compatible ▪ Suitable for fluorescence assay, luminescence assay ▪ TC Treated for cell culture & adherent cell types ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Alphanumeric labeling ▪ Homogeneous cell culture ▪ Flat bottom ▪ USP guideline compliant (USP class Ⅵ tested) ▪ ANSI guideline compliant
Catalog No. 50-233-5969 Supplier Alkali Scientific Supplier No. TPW9096
Add to Cart
Add to Cart
White plates for luminescence assays provide maximum reflection and help minimize cross-talk. The entire body and flat surface of this plate is homogenious white color. Provided with a compatible, low evaporation lid. ▪ HTS (High-Throughput Screening) compatible ▪ Suitable for fluorescence assay, luminescence assay ▪ TC Treated for cell culture & adherent cell types ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Alphanumeric labeling ▪ Homogeneous cell culture ▪ Flat bottom ▪ USP guideline compliant (USP class Ⅵ tested) ▪ ANSI guideline compliant