Learn More
Electron Microscopy Sciences MultiDish 4 Well Rectangle, 24 x 67 mm wells, Sterile, 3.5-8.0 mL Working volume
SDP
Catalog No. 501899311
Encompass_Preferred
MultiDish Surface is useful in all areas of cell culture including scale-up and cloning. Raised well rims to lower risk of cross contamination. Excellent optical quality. Nunclon™Δ certified.
Well size: 24 x 67 mm
Working volume: 3.5-8.0 mL
Area: 16 cm2
Plate size: 85.5x127.8 mm
Catalog No. 50-189-9311 Supplier Electron Microscopy Sciences Supplier No. 70440100
Add to Cart
Add to Cart
MultiDish Surface is useful in all areas of cell culture including scale-up and cloning. Raised well rims to lower risk of cross contamination. Excellent optical quality. Nunclon™Δ certified.
Well size: 24 x 67 mm
Working volume: 3.5-8.0 mL
Area: 16 cm2
Plate size: 85.5x127.8 mm