Learn More
Electron Microscopy Sciences Nunclon™ Multidish 6-Well, Round Poly-D-Lysine
SDP
Catalog No. NC3661914
Encompass_Preferred
Useful in all areas of cell culture including scale-up and cloning. Raised well rims to lower risk of cross contamination. Excellent optical quality. Nunclon™ certified. Multidish 6 is available coated with Poly-D-Lysine or Collagen.
Catalog No. NC3661914 Supplier Electron Microscopy Sciences Supplier No. 70444
Useful in all areas of cell culture including scale-up and cloning. Raised well rims to lower risk of cross contamination. Excellent optical quality. Nunclon™ certified. Multidish 6 is available coated with Poly-D-Lysine or Collagen.