Learn More
CHEMetrics™ by AquaPhoenix Multi-analyte Photometer for Water Quality Analysis
SDP
Catalog No. 13299712
Change view
Click to view available options
Type:
Multi-Analyte Photometer
Catalog No. Type
13-299-712 Multi-Analyte Photometer
1 options
Catalog No. 13-299-712 Supplier CHEMetrics™ by AquaPhoenix Supplier No. V3000
Add to Cart
Add to Cart
Portable multi-analyte water photometer for field and laboratory applications. Features an intuitive menu and over 40 pre-programmed calibrations for Vacu-vials ampoules and MDL test kits.
  • Field portable Rechargeable battery and universal power pack (Power LabStation required) Waterproof Web-based methods update Data logging storage for 100 data sets Language selections: English, German, French, Spanish
Specifications
pH Range 7
Type Multi-Analyte Photometer