Learn More
Electron Microscopy Sciences Multi-Angle Swivel Vacu-vise, 1 ea
SDP
Catalog No. 50996496
Encompass_Preferred
With a rubber suction base; you just flick off the Vacu-lever and the Vacu-vise firmly fastens to any clean, smooth surface. When you lift the Vacu-lever up , the Vacu-vise is released. Multi-Angle Swivel Vacu-vise can be tilted on any desired angle. Swivels 360 degrees. 3" metal jaws and it is supplied with a slide-on rubber to hold delicate objects; opens up to 2 5/8"; 6 1/4" triangle base.
Catalog No. 50-996-496 Supplier Electron Microscopy Sciences Supplier No. 62068
With a rubber suction base; you just flick off the Vacu-lever and the Vacu-vise firmly fastens to any clean, smooth surface. When you lift the Vacu-lever up , the Vacu-vise is released. Multi-Angle Swivel Vacu-vise can be tilted on any desired angle. Swivels 360 degrees. 3" metal jaws and it is supplied with a slide-on rubber to hold delicate objects; opens up to 2 5/8"; 6 1/4" triangle base.