Learn More
Euthanex Multi-Animal Breather for Anesthesia System with 5-ports
Catalog No. 501540151
Encompass_Preferred
Multi-Animal Breather with 5-ports for up to five mice or three rats. Single gas line supplies the multiple outlets. Exhaust line runs to a charcoal filter for safe gas evac. Includes (5) EZ-110 mouse nosecones and (3) EZ-112 rat nosecones.
Catalog No. 50-154-0151 Supplier Euthanex Supplier No. EZ109
Multi-Animal Breather with 5-ports for up to five mice or three rats. Single gas line supplies the multiple outlets. Exhaust line runs to a charcoal filter for safe gas evac. Includes (5) EZ-110 mouse nosecones and (3) EZ-112 rat nosecones.