Learn More
Multi Anion Standard 2, For IC, in water, MilliporeSigma™ Supelco™
Catalog No. 111004236 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
11-100-4236 100 mL
1 options
Catalog No. 11-100-4236 Supplier MilliporeSigma Supelco Supplier No. 53798100ML
Add to Cart
Add to Cart
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
TRUSTED_SUSTAINABILITY
Specifications
Product Line TraceCERT
Quantity 100 mL
Grade TraceCERT™ Analytical Standard
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC