Learn More
Meso Scale Discovery 96-well Small Spot Goat Anti-Mouse (GAM) SECTOR Plate, Standard surface, For Research Use Only, 2-8 °C storage, Compatible with MSD SECTOR Imagers, Minimum order size 25 plates
SDP
Catalog No. NC2540836
Encompass
The MESO SCALE DISCOVERY MULTI-ARRAY 96-well Small Spot Goat Anti-Mouse (GAM) SECTOR Plate is designed for sensitive and precise detection in various research applications. Featuring a standard surface coated with Goat Anti-Mouse antibody, this plate ensures compatibility with multiple MSD SECTOR Imagers and QuickPlex instruments, providing enhanced sensitivity and reduced non-specific binding in complex biological samples.
  • High sensitivity for enhanced signal detection
  • Reduced non-specific binding for accurate results
  • Compatible with numerous MSD imaging and detection instruments
  • Optimized for rapid immunoassay development
Catalog No. NC2540836 Supplier Meso Scale Discovery Supplier No. L45MA1
The MESO SCALE DISCOVERY MULTI-ARRAY 96-well Small Spot Goat Anti-Mouse (GAM) SECTOR Plate is designed for sensitive and precise detection in various research applications. Featuring a standard surface coated with Goat Anti-Mouse antibody, this plate ensures compatibility with multiple MSD SECTOR Imagers and QuickPlex instruments, providing enhanced sensitivity and reduced non-specific binding in complex biological samples.
  • High sensitivity for enhanced signal detection
  • Reduced non-specific binding for accurate results
  • Compatible with numerous MSD imaging and detection instruments
  • Optimized for rapid immunoassay development