Learn More
Mopec Multi-barrier Pouch
SDP
Catalog No. 22446121 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
3.7 x 3 in.
4.4 x 6.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-121 3.7 x 3 in.
22-446-092 4.4 x 6.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-121 Supplier Mopec Supplier No. FS161
Add to Cart
Add to Cart
Designed to protect cards from exposure to gases and liquids while supporting secure storage and labeling.
  • Outer paper layer can be used for labeling or writing
  • Maintains sample security
  • Available in small and large sizes.
  • Designed to protect FTA™ cards from exposure to gases and liquids.
  • The Multi-barrier Pouch is a tamper evident, seven layer laminated pouch designed
Specifications
Dimensions (L x W) 3.7 x 3 in.
Product Type Multi-barrier Pouch
Color Gray
Quantity 100/Pk.