Learn More
Contec Multi Bucket System with Flat Mops
Catalog No. 19991016 Shop All Contec Inc. Products
Change view
Click to view available options
Material:
Polypropylene Bucket, Stainless Steel Cart
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-991-016 Stainless Steel
19-991-014 Polypropylene Bucket, Stainless Steel Cart
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-991-016 Supplier Contec Supplier No. 2707U2SF
Add to Cart
Add to Cart
Contec's Multi-Bucket System offers flexibility with two or three buckets in stainless steel or polypropylene.
The Contec Multi-Bucket System is designed for use with Contec's flat mops, offering flexibility, mobility, and easy customization for cleanroom cleaning.
  • Configurable with two or three buckets in stainless steel or polypropylene
  • Extendable front frame accommodates an additional bucket for added capacity
  • Quiet, locking casters ensure smooth movement and stability
  • Utility handle for easy transport of cleaning supplies
Hands-free mop removal tool available for VertiKlean™ MAX™ and QuickTask™ mop heads
  • Integrated mop holder with brace for secure storage during transport
  • Compatible with stainless steel downpress wringer for flat mops
  • Autoclavable at 121°C for 30 minutes for sterilization in critical environments
Specifications
Product Type 2 Bucket System
Material Stainless Steel