Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ Multi-Channel Micropipette -12 Transferpette™ pro, Adjustable
GREENER_CHOICE

Catalog No. 13200193 Shop All BrandTech Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
50 μL
100 μL
200 μL
300 μL
1250 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-200-193 10 μL
13-200-194 50 μL
13-200-183 100 μL
13-200-198 200 μL
13-200-171 300 μL
13-200-188 1250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-200-193 Supplier BRANDTECH™ Supplier No. 706920
Add to Cart
Add to Cart

Slim multi-channel micropipette with adjustable finger rest for ergonomic use, flexible calibration options, and built-in counter protection for durability.

  • Its slim design and adjustable finger rest ensure that the pipette fits perfectly in your hand, making it ergonomic and comfortable to use, even during long pipetting sessions
  • To keep you flexible at all times, the Transferpette™ pro features two adjustment techniques:
    • Easy calibration for permanent calibration without tools
    • User adjustment for quick, reversible adaptation to changing conditions such as temperature, tip shape, or media properties
  • The Transferpette™ pro is made of high-quality materials with built-in counter protection, making it both robust and durable. It's fully autoclavable (121°C) and available as single- and multi-channel pipettes.

Limitations of use:

  • Operating temperature should be between +15°C and +40°C (59°F to 104°F). Other temperatures upon request.
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa·s
  • For viscous media, the speed must be adjusted if necessary
TRUSTED_SUSTAINABILITY

Specifications

Accuracy < ±1.6%
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Gray
Disposable Reusable
For Use With Pipette Tip 0.5 to 20 μL
Grip Type Ergonomic
Material Polypropylene
Model Transferpette Pro
Number of Channels 12
Programmability No
Sterility Non-sterile
Type Mechanical Pipette
Volume (Metric) 10 μL
Includes Performance Certificate, 2 x TipBox Filled with BRAND™Pipette Tips, Shelf Mount, Reagent Reservoir, Installation Tool, Silicone Grease, Set of Sealing Rings
Increments 0.01 μL
Precision < 1% (CV)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.