Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Multi-Channel Pipettor and Pipette Aid Holder
SDP

Catalog No. 01259216
Change view
Click to view available options
Mounting:
Magnet Mount
Tape Mount
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-216 Tape Mount
01-259-187 Magnet Mount
01-259-189 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-216 Supplier Poltex Supplier No. MULPIP421T
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A

Holds 2 multichannel pipettes, 4 single channel pipettes, and 1 pipette aid. Choose from Wall Mount, HB Tape Mount, or Magnet Mount with durable PETG construction.

  • The Multi Pipette Hanger holds 2 multichannel, 4 single channel, and 1 pipette filler or pipette aid.
  • Choose from Wall Mount (screws/anchors included), HB Tape Mount (high-bond tape included), or Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Single- and Multi-channel Pipettes, Single-channel or Multi-channel Pipetter, Pipettes, Pipette Aid
Material PETG
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Application) Multi Pipette Hanger
Mounting Tape Mount
Dimensions (L x W x H) 3.5 x 14.5 x 3.7 in.
Weight (Metric) 1 lb.
Electrical Requirements None
Includes High Bond Tape for Mounting
Product Type Pipette Holder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.