Learn More
Advansta Inc Multi-chamber incubation trays, 2/pk
Catalog No. NC3235499
Encompass
These lidded incubation trays have a perfectly smooth interior surface, making them ideal for staining and washing electrophoresis gels and membranes. Trays are manufactured to prevent adsorption of antibodies.
Catalog No. NC3235499 Supplier Advansta Inc Supplier No. L07080002
These lidded incubation trays have a perfectly smooth interior surface, making them ideal for staining and washing electrophoresis gels and membranes. Trays are manufactured to prevent adsorption of antibodies.