Learn More
MSC Minuteman Degreaser, Tough Green, Environmental, 5 Gallon Bag-in-Box
Catalog No. 502839416
Encompass
This environmental degreaser is designed for powerful cleaning that is safe for both the user and the environment. Its unique blend of biodegradable surfactants and environmentally safe solvents effectively penetrates and suspends dirt, grease, and grime. Suitable for a wide range of water-safe surfaces and applications, this versatile product is ideal for various institutional and industrial settings.
  • Powerful, safe, and environmentally friendly degreasing
  • Biodegradable and phosphate-free formula
  • Non-toxic, non-corrosive, and not a skin sensitizer
  • Meets VOC requirements and has low aquatic toxicity
  • Versatile for various surfaces and settings including kitchens and warehouses
  • Neutral pH (7-8) with excellent detergency
  • Meets USDA C-1 type product standards and EPA safer chemistry recognition
Catalog No. 50-283-9416 Supplier MSC Supplier No. 57184723
This environmental degreaser is designed for powerful cleaning that is safe for both the user and the environment. Its unique blend of biodegradable surfactants and environmentally safe solvents effectively penetrates and suspends dirt, grease, and grime. Suitable for a wide range of water-safe surfaces and applications, this versatile product is ideal for various institutional and industrial settings.
  • Powerful, safe, and environmentally friendly degreasing
  • Biodegradable and phosphate-free formula
  • Non-toxic, non-corrosive, and not a skin sensitizer
  • Meets VOC requirements and has low aquatic toxicity
  • Versatile for various surfaces and settings including kitchens and warehouses
  • Neutral pH (7-8) with excellent detergency
  • Meets USDA C-1 type product standards and EPA safer chemistry recognition