Learn More
MSC Zep Cleaner and Degreaser, Concentrated, Green Seal Certified
Supplier:  MSC 40249930
Encompass
Multi-Clean Green is a concentrated, heavy-duty, multi-purpose cleaner and degreaser. It is formulated to be non-carcinogenic, non-toxic, RCRA non-hazardous, and free of chlorinated solvents. This Green Seal Certified product effectively cleans a wide variety of surfaces including ovens, grills, machinery, walls, floors, and countertops, making it suitable for industrial, institutional, and food service applications.
  • Green Seal™ certified for industrial and institutional use.
  • RCRA non-hazardous, meets defined requirements.
  • Economical, dilutes for various cleaning levels.
  • Versatile, eliminates need for multiple cleaning products.
  • Effectively cleans and degreases surfaces, rinses completely.
  • Non-flammable, free of petroleum or chlorinated solvents.
  • Contains no added fragrance.
  • Biodegradable surfactants.
  • Suitable for many applications and surfaces.
Catalog No. 50-283-9025