Learn More
MSC Multi-Clean Glass and Surface Cleaner, Unscented, 2 L, 4/Case
Catalog No. 502867854
Encompass
Multi-Shine Glass & Surface Cleaner is an effective blend of surfactants designed to penetrate and lift surface films for easy, streak-free cleaning. This multi-purpose cleaner is suitable for glass, mirrors, and various polished or painted surfaces. It is formulated without ammonia, alcohol, or petroleum-derived solvents, and meets Green Seal™ Standard GS-37.
  • Effectively cuts dirt and oil
  • Multi-purpose for glass, mirrors, and polished/painted surfaces including brass, aluminum, vinyl, plastics, Formica, and stainless steel
  • Leaves surfaces sparkling clean and streak-free with minimal effort
  • Ammonia, alcohol, and petroleum-derived solvent-free for safer use
  • Green Seal certified for environmental sustainability
Catalog No. 50-286-7854 Supplier MSC Supplier No. 68906916
Multi-Shine Glass & Surface Cleaner is an effective blend of surfactants designed to penetrate and lift surface films for easy, streak-free cleaning. This multi-purpose cleaner is suitable for glass, mirrors, and various polished or painted surfaces. It is formulated without ammonia, alcohol, or petroleum-derived solvents, and meets Green Seal™ Standard GS-37.
  • Effectively cuts dirt and oil
  • Multi-purpose for glass, mirrors, and polished/painted surfaces including brass, aluminum, vinyl, plastics, Formica, and stainless steel
  • Leaves surfaces sparkling clean and streak-free with minimal effort
  • Ammonia, alcohol, and petroleum-derived solvent-free for safer use
  • Green Seal certified for environmental sustainability