Learn More
Electron Microscopy Sciences Multi-colored Well Orienter for 96 Well Microplates
SDP
Supplier:  Electron Microscopy Sciences 7049018
Encompass_Preferred
Bright colors make it easy todistinguish between columnsand rows. Fits most standard flat andround bottom 96-well and 384-well plates. Ideal for use with The Pipetter Guide for efficient and stress-free identification of rows and columns.
Catalog No. 50-930-337