Learn More
Perkin Elmer US LLC 29-Element Solution, 5% HNO3, 10 µg/mL, 100 mL
Catalog No. 509045835
Encompass_Preferred
Details Concentration Element 10 µg/mL Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn * Supplied in a separate bottle
Catalog No. 50-904-5835 Supplier Perkin Elmer US LLC Supplier No. N9301720
Details Concentration Element 10 µg/mL Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn * Supplied in a separate bottle