Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Multi-Element ICH/USP Oral Standard Solution A, Specpure™

Catalog No. AA47318AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47318-AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA47318-AE Supplier Thermo Scientific™ Supplier No. 47318AE
Add to Cart

missing translation for 'validMainframeMsg'

Form: Liquid | Matrix: 2% HNO{3}

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Molecular Formula Matrix: 2% HNO{3}
Concentration Hg 30 μg/mL ++2%
UN Number UN3264
Boiling Point 100°C
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.