Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multi-Element Organometallic Oil Standards 23 Element, 300μg/g; SPEX CertiPrep™
SDP

Catalog No. S23300Z Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S23300Z 50 g
S23300Y 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S23300Z Supplier SPEX CertiPrep Supplier No. S23300Z
Add to Cart
Add to Cart

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag300μg/g
Al300μg/g
B300μg/g
Ba300μg/g
Ca300μg/g
Cd300μg/g
Cr300μg/g
Cu300μg/g
Fe300μg/g
K300μg/g
Mg300μg/g
Mn300μg/g
Mo300μg/g
Na300μg/g
Ni300μg/g
P300μg/g
Pb300μg/g
Sb300μg/g
Si300μg/g
Sn300μg/g
Ti300μg/g
V300μg/g
Zn300μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material 23 Element Oil Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Al,Ba,B
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Al, Ba, B
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Quantity 50 g
WARNING: Cancer - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.