Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multi-Element Plasma Standard Solution 2, Specpure™

Catalog No. AA42882AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42882AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42882AE Supplier Thermo Scientific Chemicals Supplier No. 042882AE
Add to Cart

missing translation for 'validMainframeMsg'

 

Multi-Element Plasma Standard Solution is used in atomic emission spectrometry, mass spectrometry and also in biomedical samples. It is also used to prepare calibration and standard solutions for plasma emission spectrometry. Further, it is widely used analytical technique for trace metal analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Multi-Element Plasma Standard Solution 2
Physical Form Liquid
MDL Number MFCD00212338
UN Number UN3264
Solubility Miscible with water
Quantity 100 mL
Grade Specpure

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.