Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multi-Element QC-21 Check Standard Solution, Specpure™

Catalog No. AA41328AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41328AP 500 mL
AA41328AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41328AP Supplier Thermo Scientific Chemicals Supplier No. 041328AP
Add to Cart

missing translation for 'validMainframeMsg'

Multi-Element QC-21 Check Standard Solution is used as a certified reference material or quality control check standard for inductively coupled plasma optical emission spectroscopy (ICP-OES). It is also used in inductively coupled plasma mass spectrometry (ICP-MS), and flame or furnace atomic absorption spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Multi-Element QC-21 Check Standard Solution
Physical Form Liquid
Concentration Matrix: 5% HNO3/tr. F-/tr. tartaric acid
MDL Number MFCD00803610
UN Number UN3264
Solubility Miscible with water
Type Analytical Standard
Quantity 500 mL
Grade Specpure

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.