Learn More
CELLTREAT Scientific Products Lab Grab Multi-Use Extension Grabber, 2/CS
SDP
Catalog No. 501535078
Encompass_Preferred
CELLTREAT Scientific Products Lab Grab. Allows hands-free and easy grasping of small items, as well as hot or cold samples. Large extension radius to pick up small to mid-sized objects including cryogenic vials, screw top micro tubes, micro centrifuge tubes, and cell strainers. Retraction feature aids in stability and strength of grip. Textured handle prevents slipping when wearing gloves. Non-sterile. Packed 5 per bag.
Catalog No. 50-153-5078 Supplier CELLTREAT Scientific Products Supplier No. 230110
CELLTREAT Scientific Products Lab Grab. Allows hands-free and easy grasping of small items, as well as hot or cold samples. Large extension radius to pick up small to mid-sized objects including cryogenic vials, screw top micro tubes, micro centrifuge tubes, and cell strainers. Retraction feature aids in stability and strength of grip. Textured handle prevents slipping when wearing gloves. Non-sterile. Packed 5 per bag.